1-Bromo-4-chloro-2-fluoro-5-methylbenzene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-chloro-4-fluorotoluene

PC53588-1G PC53588-25G PC53588-5G
APOSPC53588-1GEA 21.9 EUR
APOSPC53588-1G APOSPC53588-25G APOSPC53588-5G
1-Bromo-4-chloro-2-fluoro-5-methylbenzene 95%
1-Bromo-4-chloro-2-fluoro-5-methylbenzene
Formel: C₇H₅BrClF
Molecular Weight: 223,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800685
CAS-Nummer: 200190-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR