Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate 95%

Lieferant: Apollo Scientific

PC53610-1G PC53610-250MG PC53610-5G
APOSPC53610-1GEA 260 EUR
APOSPC53610-1G APOSPC53610-250MG APOSPC53610-5G
Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate 95%
Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate
Formel: C₈H₄Cl₂FNO₄
Molecular Weight: 268,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12172978
CAS-Nummer: 1153285-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR