Methyl 3-chloro-4,5-difluoro-benzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-chloro-4,5-difluorobenzoate

PC53616-1G PC53616-5G
APOSPC53616-1GEA 42.2 EUR
APOSPC53616-1G APOSPC53616-5G
Methyl 3-chloro-4,5-difluoro-benzoate 95%
Methyl 3-chloro-4,5-difluoro-benzoate
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698002
CAS-Nummer: 1214344-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR