Methyl 4-fluoronaphthalene-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 4-fluoro-1-naphthoate

PC53620-1G PC53620-250MG
APOSPC53620-1GEA 348 EUR
APOSPC53620-1G APOSPC53620-250MG
Methyl 4-fluoronaphthalene-1-carboxylate
Methyl 4-fluoronaphthalene-1-carboxylate
Formel: C₁₂H₉FO₂
Molecular Weight: 204,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025244
CAS-Nummer: 13772-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR