4-(Carboxymethyl)-3-fluorobenzeneboronic acid

Lieferant: Apollo Scientific

PC53689-1G PC53689-250MG PC53689-5G
APOSPC53689-1GEA 271 EUR
APOSPC53689-1G APOSPC53689-250MG APOSPC53689-5G
4-(Carboxymethyl)-3-fluorobenzeneboronic acid
4-(Carboxymethyl)-3-fluorobenzeneboronic acid
Formel: C₈H₈BFO₄
Molecular Weight: 197,956 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32220289
CAS-Nummer: 1146614-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR