2-Bromo-3-fluoro-6-(trifluoromethyl)pyridine 98%
Lieferant: Apollo Scientific
PC5374-1G
APOSPC5374-1GEA
59.6
EUR
APOSPC5374-1G
2-Bromo-3-fluoro-6-(trifluoromethyl)pyridine 98%
2-Bromo-3-fluoro-6-(trifluoromethyl)pyridine
Formel:
C₆H₂BrF₄N Molecular Weight: 243,984 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226611 CAS-Nummer: 1159512-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...