3-(Trifluoromethylthio)benzaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-((Trifluoromethyl)thio)benzaldehyde

PC5389-10G PC5389-1G PC5389-5G
APOSPC5389-10GEA 500 EUR
APOSPC5389-10G APOSPC5389-1G APOSPC5389-5G
3-(Trifluoromethylthio)benzaldehyde 95+%
3-(Trifluoromethylthio)benzaldehyde
Formel: C₈H₅F₃OS
Lagertemperatur: Kühlschrank
MDL: MFCD00236335
CAS-Nummer: 51748-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR