(R)-3-Fluoropyrrolidine-1-sulphonyl chloride ≥98%

Lieferant: Apollo Scientific

PC540072-1G PC540072-100MG PC540072-250MG
APOSPC540072-1GEA 473 EUR
APOSPC540072-1G APOSPC540072-100MG APOSPC540072-250MG
(R)-3-Fluoropyrrolidine-1-sulphonyl chloride ≥98%
(R)-3-Fluoropyrrolidine-1-sulphonyl chloride
Formel: C₄H₇ClFNO₂S
Molecular Weight: 187,62 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30533071
CAS-Nummer: 1411774-27-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR