5-Bromo-1-chloro-2,3-difluorobenzene 99%

Lieferant: Apollo Scientific
Achtung

PC54126-1G PC54126-250MG PC54126-5G
APOSPC54126-1GEA 79.9 EUR
APOSPC54126-1G APOSPC54126-250MG APOSPC54126-5G
5-Bromo-1-chloro-2,3-difluorobenzene 99%
5-Bromo-1-chloro-2,3-difluorobenzene
Formel: C₆H₂BrClF₂
Molecular Weight: 227,434 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845915
CAS-Nummer: 1060813-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR