4-Nitro-3-(trifluoromethyl)benzenesulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
PC5419-1G
PC5419-5G
APOSPC5419-1GEA
140
EUR
APOSPC5419-1G
APOSPC5419-5G
4-Nitro-3-(trifluoromethyl)benzenesulphonamide
4-Nitro-3-(trifluoromethyl)benzenesulphonamide
Formel:
C₇H₅F₃N₂O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094293 CAS-Nummer: 21988-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...