5-Methyl-2-(trifluoromethyl)furan-3-carboxamide 97%

Lieferant: Apollo Scientific
Achtung

PC5436-1G PC5436-250MG
APOSPC5436-1GEA 364 EUR
APOSPC5436-1G APOSPC5436-250MG
5-Methyl-2-(trifluoromethyl)furan-3-carboxamide 97%
5-Methyl-2-(trifluoromethyl)furan-3-carboxamide
Formel: C₇H₆F₃NO₂
Molecular Weight: 193,123 g/mol
Schmelzpunkt: 120…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276988
CAS-Nummer: 175276-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR