3-Chloro-2,4-difluorobenzyl alcohol 98%

Lieferant: Apollo Scientific

PC5442-5G PC5442-1G
APOSPC5442-5GEA 313 EUR
APOSPC5442-5G APOSPC5442-1G
3-Chloro-2,4-difluorobenzyl alcohol 98%
3-Chloro-2,4-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116002
CAS-Nummer: 886501-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR