6-Fluoro-4-hydrazinoquinoline hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Fluoroquinolin-4-yl)hydrazine hydrochloride, 6-Fluoro-4-hydrazino-1-azanaphthalene hydrochloride

PC5461-250MG PC5461-100MG PC5461-1G
APOSPC5461-250MGEA 154 EUR
APOSPC5461-250MG APOSPC5461-100MG APOSPC5461-1G
6-Fluoro-4-hydrazinoquinoline hydrochloride
6-Fluoro-4-hydrazinoquinoline hydrochloride
Formel: C₉H₈FN₃·ClH
Molecular Weight: 213,639 g/mol
Schmelzpunkt: >280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02684141
CAS-Nummer: 1172049-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR