3-Hydroxy-4,4,4-trifluorobutyramide

Lieferant: Apollo Scientific
Achtung

PC5462-1G PC5462-5G PC5462-250MG
APOSPC5462-1GEA 94.4 EUR
APOSPC5462-1G APOSPC5462-5G APOSPC5462-250MG
3-Hydroxy-4,4,4-trifluorobutyramide
3-Hydroxy-4,4,4-trifluorobutyramide
Formel: C₄H₆F₃NO₂
Molecular Weight: 157,091 g/mol
Schmelzpunkt: 124…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784293
CAS-Nummer: 453-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR