4-(Trifluoromethylsulphonyl)-N-ethylaniline

Lieferant: Apollo Scientific

PC5467-1G PC5467-250MG
APOSPC5467-1GEA 706 EUR
APOSPC5467-1G APOSPC5467-250MG
4-(Trifluoromethylsulphonyl)-N-ethylaniline
4-(Trifluoromethylsulphonyl)-N-ethylaniline
Formel: C₉H₁₀F₃NO₂S
Molecular Weight: 253.241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631696
CAS-Nummer: 240490-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR