2-(N-Methylhydrazino)-5-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC5503-1G PC5503-250MG PC5503-5G
APOSPC5503-1GEA 58 EUR
APOSPC5503-1G APOSPC5503-250MG APOSPC5503-5G
2-(N-Methylhydrazino)-5-(trifluoromethyl)pyridine 97%
2-(N-Methylhydrazino)-5-(trifluoromethyl)pyridine
Formel: C₇H₈F₃N₃
Schmelzpunkt: 52…55 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052297
CAS-Nummer: 163620-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR