6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC55148-1G
PC55148-250MG
APOSPC55148-1GEA
696
EUR
APOSPC55148-1G
APOSPC55148-250MG
6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline 95%
6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinoline
Formel:
C₁₀H₉BrF₃NO Molecular Weight: 296,084 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922517 CAS-Nummer: 2366994-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...