Pentafluorobenzyl acrylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Pentafluorobenzyl prop-2-enoate

PC5537-1G PC5537-5G PC5537-250MG
APOSPC5537-1GEA 198 EUR
APOSPC5537-1G APOSPC5537-5G APOSPC5537-250MG
Pentafluorobenzyl acrylate 97%
Pentafluorobenzyl acrylate
Formel: C₁₀H₅F₅O₂
Molecular Weight: 252,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042328
CAS-Nummer: 153614-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR