4-Bromo-2-fluoro-1-(3,3,3-trifluoroprop-1-ynyl)benzene

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-3-Fluoro-4-(3,3,3-trifluoro-1-propyn-1-yl)benzene

PC55374-1G PC55374-250MG
APOSPC55374-1GEA 654 EUR
APOSPC55374-1G APOSPC55374-250MG
4-Bromo-2-fluoro-1-(3,3,3-trifluoroprop-1-ynyl)benzene
4-Bromo-2-fluoro-1-(3,3,3-trifluoroprop-1-ynyl)benzene
Formel: C₉H₃BrF₄
Molecular Weight: 267.018 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 2137951-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR