2-(Pentafluoropropenyl)pivalate 98%
Lieferant: Apollo Scientific
Synonyme:
2,2-Difluoro-1-(trifluoromethyl)vinyl pivalate
PC5577-25G
PC5577-5G
PC5577-1G
APOSPC5577-25GEA
830
EUR
APOSPC5577-25G
APOSPC5577-5G
APOSPC5577-1G
2-(Pentafluoropropenyl)pivalate 98%
2-(Pentafluoropropenyl)pivalate
Formel:
C₈H₉F₅O₂ Molecular Weight: 232,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038873 CAS-Nummer: 885275-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...