2,6-Difluoro-4-methylbenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC56022-1G PC56022-5G
APOSPC56022-1GEA 66.7 EUR
APOSPC56022-1G APOSPC56022-5G
2,6-Difluoro-4-methylbenzoic acid 98%
2,6-Difluoro-4-methylbenzoic acid
Formel: C₈H₆F₂O₂
Molecular Weight: 172.129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847292
CAS-Nummer: 1201597-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR