Methyl 3-fluoro-4-(methylsulphonyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC56035-1G PC56035-5G PC56035-25G
APOSPC56035-1GEA 111 EUR
APOSPC56035-1G APOSPC56035-5G APOSPC56035-25G
Methyl 3-fluoro-4-(methylsulphonyl)benzoate
Methyl 3-fluoro-4-(methylsulphonyl)benzoate
Formel: C₉H₉FO₄S
Molecular Weight: 232,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143295
CAS-Nummer: 1215074-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR