4-Bromo-2-fluoro-6-methylaniline

Lieferant: Apollo Scientific
Achtung

PC56036-1G PC56036-5G
APOSPC56036-1GEA 59.9 EUR
APOSPC56036-1G APOSPC56036-5G
4-Bromo-2-fluoro-6-methylaniline
4-Bromo-2-fluoro-6-methylaniline
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22683228
CAS-Nummer: 429683-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR