3-(Trifluoromethylsulphonyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-3-(trifluoromethylsulphonyl)benzene
PC56048-1G
APOSPC56048-1GEA
955
EUR
APOSPC56048-1G
3-(Trifluoromethylsulphonyl)benzyl bromide
3-(Trifluoromethylsulphonyl)benzyl bromide
Formel:
C₈H₆BrF₃O₂S Molecular Weight: 303,096 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706281 CAS-Nummer: 866208-07-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...