2,2,2,2',6'-Pentafluoro-4'-(trifluoromethyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[2,6-Difluoro-4-(trifluoromethyl)phenyl]-2,2,2-trifluoroethanone

PC56058-1G PC56058-250MG PC56058-5G
APOSPC56058-1GEA 1320 EUR
APOSPC56058-1G APOSPC56058-250MG APOSPC56058-5G
2,2,2,2',6'-Pentafluoro-4'-(trifluoromethyl)acetophenone
2,2,2,2',6'-Pentafluoro-4'-(trifluoromethyl)acetophenone
Formel: C₉H₂F₈O
Molecular Weight: 278,099 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30746954
CAS-Nummer: 2149590-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR