N-Methylperfluorobutanesulphonamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1,2,2,3,3,4,4,4-nonafluoro-N-methylbutane-1-sulphonamide, N-Methylnonafluorobutane-1-sulphonamide
PC56060-1G
PC56060-250MG
PC56060-5G
PC56060-100MG
APOSPC56060-1GEA
516
EUR
APOSPC56060-1G
APOSPC56060-250MG
APOSPC56060-5G
APOSPC56060-100MG
N-Methylperfluorobutanesulphonamide 97%
N-Methylperfluorobutanesulphonamide
Formel:
C₅H₄F₉NO₂S Molecular Weight: 313,141 g/mol Schmelzpunkt: 36…37 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19707291 CAS-Nummer: 68298-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...