3-(Trifluoromethylsulphonyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC56069-1G PC56069-250MG
APOSPC56069-1GEA 671 EUR
APOSPC56069-1G APOSPC56069-250MG
3-(Trifluoromethylsulphonyl)benzaldehyde
3-(Trifluoromethylsulphonyl)benzaldehyde
Formel: C₈H₅F₃O₃S
Molecular Weight: 238,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25460320
CAS-Nummer: 1274904-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR