4'-Bromo-2,2,2,2',5'-pentafluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2,5-difluorophenyl)-2,2,2-trifluoroethanone

PC56078-1G PC56078-250MG
APOSPC56078-1GEA 546 EUR
APOSPC56078-1G APOSPC56078-250MG
4'-Bromo-2,2,2,2',5'-pentafluoroacetophenone
4'-Bromo-2,2,2,2',5'-pentafluoroacetophenone
Formel: C₈H₂BrF₅O
Molecular Weight: 288.997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24107308
CAS-Nummer: 1540160-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR