3-[(Trifluoromethyl)sulphonyl]phenacyl bromide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-[3-(trifluoromethylsulphonyl)phenyl]ethanone

PC56120-1G PC56120-250MG
APOSPC56120-1GEA 766 EUR
APOSPC56120-1G APOSPC56120-250MG
3-[(Trifluoromethyl)sulphonyl]phenacyl bromide
3-[(Trifluoromethyl)sulphonyl]phenacyl bromide
Formel: C₉H₆BrF₃O₃S
Molecular Weight: 331.106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536707
CAS-Nummer: 2091023-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR