2-Bromo-4-fluoro-3-methyl-6-nitroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-1-fluoro-4-methoxy-2-methyl-5-nitrobenzene

PC56128-1G PC56128-5G
APOSPC56128-1GEA 139 EUR
APOSPC56128-1G APOSPC56128-5G
2-Bromo-4-fluoro-3-methyl-6-nitroanisole
2-Bromo-4-fluoro-3-methyl-6-nitroanisole
Formel: C₈H₇BrFNO₃
Molecular Weight: 264,048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31543788
CAS-Nummer: 2187435-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR