3-Bromo-5-fluoro-2-methoxy-4-methylaniline

Lieferant: Apollo Scientific
Achtung

PC56132-1G PC56132-250MG
APOSPC56132-1GEA 473 EUR
APOSPC56132-1G APOSPC56132-250MG
3-Bromo-5-fluoro-2-methoxy-4-methylaniline
3-Bromo-5-fluoro-2-methoxy-4-methylaniline
Formel: C₈H₉BrFNO
Molecular Weight: 234.066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31543789
CAS-Nummer: 2187434-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR