6-(Trifluoromethoxy)-1H-indazole 95%

Lieferant: Apollo Scientific
Achtung

PC56142-1G PC56142-250MG
APOSPC56142-1GEA 654 EUR
APOSPC56142-1G APOSPC56142-250MG
6-(Trifluoromethoxy)-1H-indazole 95%
6-(Trifluoromethoxy)-1H-indazole
Formel: C₈H₅F₃N₂O
Molecular Weight: 202,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27993300
CAS-Nummer: 105391-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR