6-Bromo-5-fluoronicotinonitrile 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-fluoropyridine-3-carbonitrile

PC56146-1G PC56146-250MG
APOSPC56146-1GEA 641 EUR
APOSPC56146-1G APOSPC56146-250MG
6-Bromo-5-fluoronicotinonitrile 95+%
6-Bromo-5-fluoronicotinonitrile
Formel: C₆H₂BrFN₂
Molecular Weight: 200,996 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28735373
CAS-Nummer: 1806062-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR