Potassium (2-acetoxyethyl)trifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium, 2-acetyloxyethyl(trifluoro)boranuide
PC56180-1G
PC56180-250MG
APOSPC56180-1GEA
720
EUR
APOSPC56180-1G
APOSPC56180-250MG
Potassium (2-acetoxyethyl)trifluoroborate
Potassium (2-acetoxyethyl)trifluoroborate
Formel:
C₄H₇BF₃O₂·K Molecular Weight: 194,002 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22684703 CAS-Nummer: 1408168-77-3 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...