1-(Trifluoromethyl)ethenylboronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4,5,5-Tetramethyl-2-(3,3,3-trifluoroprop-1-en-2-yl)-1,3,2-dioxaborolane
PC56182-100MG
PC56182-250MG
APOSPC56182-100MGEA
691
EUR
APOSPC56182-100MG
APOSPC56182-250MG
1-(Trifluoromethyl)ethenylboronic acid, pinacol ester
1-(Trifluoromethyl)ethenylboronic acid, pinacol ester
Formel:
C₉H₁₄BF₃O₂ Molecular Weight: 222,012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16996395 CAS-Nummer: 1055881-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...