2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC56187-1G
PC56187-250MG
APOSPC56187-1GEA
944
EUR
APOSPC56187-1G
APOSPC56187-250MG
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
Formel:
C₈H₂BrF₄N Molecular Weight: 268,006 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31568568 CAS-Nummer: 2091606-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...