2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(3-(Trifluoromethyl)phenyl)thiazol-2-amine

PC5619-1G PC5619-500MG
APOSPC5619-1GEA 29 EUR
APOSPC5619-1G APOSPC5619-500MG
2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole 97%
2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole
Formel: C₁₀H₇F₃N₂S
Molecular Weight: 244.236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03034765
CAS-Nummer: 172848-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR