Tris(1H,1H,9H-perfluorononyl)borate

Lieferant: Apollo Scientific
Achtung

PC56212-1G PC56212-250MG
APOSPC56212-1GEA 145 EUR
APOSPC56212-1G APOSPC56212-250MG
Tris(1H,1H,9H-perfluorononyl)borate
Tris(1H,1H,9H-perfluorononyl)borate
Formel: C₂₇H₉BF₄₈O₃
Molecular Weight: 1304.093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28137328
CAS-Nummer: 1814-40-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR