Trifluoro(pyridine)boron 97%

Lieferant: Apollo Scientific
Gefahr

PC56215-1G PC56215-250MG
APOSPC56215-1GEA 430 EUR
APOSPC56215-1G APOSPC56215-250MG
Trifluoro(pyridine)boron 97%
Trifluoro(pyridine)boron
Formel: C₅H₅BF₃N
Molecular Weight: 146,906 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00971931
CAS-Nummer: 407-76-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR