4-(Hydroxymethyl)-2-(trifluoromethyl)phenol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Hydroxymethyl)-2-(trifluoromethyl)phenol

PC56257-1G PC56257-250MG
APOSPC56257-1GEA 1060 EUR
APOSPC56257-1G APOSPC56257-250MG
4-(Hydroxymethyl)-2-(trifluoromethyl)phenol 95%
4-(Hydroxymethyl)-2-(trifluoromethyl)phenol
Formel: C₈H₇F₃O₂
Molecular Weight: 192,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202982
CAS-Nummer: 773871-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR