2-Bromo-3,6-difluoroiodobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1,4-difluoro-3-iodobenzene
PC56261-1G
PC56261-250MG
APOSPC56261-1GEA
87
EUR
APOSPC56261-1G
APOSPC56261-250MG
2-Bromo-3,6-difluoroiodobenzene 95%
2-Bromo-3,6-difluoroiodobenzene
Formel:
C₆H₂BrF₂I Molecular Weight: 318.885 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12922621 CAS-Nummer: 1208074-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...