2,3-Difluoro-4-(trifluoromethyl)benzylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: [2,3-Difluoro-4-(trifluoromethyl)phenyl]methanamine

PC56282-1G PC56282-250MG PC56282-5G
APOSPC56282-1GEA 422 EUR
APOSPC56282-1G APOSPC56282-250MG APOSPC56282-5G
2,3-Difluoro-4-(trifluoromethyl)benzylamine 95%
2,3-Difluoro-4-(trifluoromethyl)benzylamine
Formel: C₈H₆F₅N
Molecular Weight: 211.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06212917
CAS-Nummer: 581813-21-0
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR