2-Nitro-4-(trifluoromethoxy)toluene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-2-nitro-4-(trifluoromethoxy)benzene
PC56284-1G
PC56284-5G
APOSPC56284-1GEA
95.7
EUR
APOSPC56284-1G
APOSPC56284-5G
2-Nitro-4-(trifluoromethoxy)toluene 95%
2-Nitro-4-(trifluoromethoxy)toluene
Formel:
C₈H₆F₃NO₃ Molecular Weight: 221,133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954767 CAS-Nummer: 70692-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...