6-Fluoro-1H-pyrrolo[3,2-b]pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC56323-1G PC56323-250MG
APOSPC56323-1GEA 193 EUR
APOSPC56323-1G APOSPC56323-250MG
6-Fluoro-1H-pyrrolo[3,2-b]pyridine 97%
6-Fluoro-1H-pyrrolo[3,2-b]pyridine
Formel: C₇H₅FN₂
Molecular Weight: 136.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12962608
CAS-Nummer: 1190320-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR