2,3-Dibromo-1,4-difluorobenzene 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC56328-1G
PC56328-25G
PC56328-5G
APOSPC56328-1GEA
21.9
EUR
APOSPC56328-1G
APOSPC56328-25G
APOSPC56328-5G
2,3-Dibromo-1,4-difluorobenzene 95+%
2,3-Dibromo-1,4-difluorobenzene
Formel:
C₆H₂Br₂F₂ Molecular Weight: 271,885 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922620 CAS-Nummer: 179737-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...