3-Bromo-2,5-difluoroiodobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2,5-difluoro-3-iodobenzene
PC56329-1G
PC56329-250MG
PC56329-5G
APOSPC56329-1GEA
509
EUR
APOSPC56329-1G
APOSPC56329-250MG
APOSPC56329-5G
3-Bromo-2,5-difluoroiodobenzene 95%
3-Bromo-2,5-difluoroiodobenzene
Formel:
C₆H₂BrF₂I Lagertemperatur: Kühlschrank |
MDL:
MFCD28737933 CAS-Nummer: 1807026-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...