1,4-Dibromo-2,5-bis(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

Synonyme: 1,4-Bis(trifluoromethyl)-2,5-dibromobenzene, 1,4-Dibromo-2,5-bis(trifluoromethyl)benzene

PC56680-1G PC56680-250MG PC56680-5G
APOSPC56680-1GEA 48 EUR
APOSPC56680-1G APOSPC56680-250MG APOSPC56680-5G
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene 95%
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene
Formel: C₈H₂Br₂F₆
Molecular Weight: 371,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526888
CAS-Nummer: 2375-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR