4-Bromo-2,5-bis(trifluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
PC56681-1G
PC56681-250MG
APOSPC56681-1GEA
862
EUR
APOSPC56681-1G
APOSPC56681-250MG
4-Bromo-2,5-bis(trifluoromethyl)benzaldehyde 95%
4-Bromo-2,5-bis(trifluoromethyl)benzaldehyde
Formel:
C₉H₃BrF₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD15526889 CAS-Nummer: 1260743-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...