Methyl 4-bromo-2,5-bis(trifluoromethyl)benzoate 95%

Lieferant: Apollo Scientific

PC56689-1G PC56689-250MG
APOSPC56689-1GEA 877 EUR
APOSPC56689-1G APOSPC56689-250MG
Methyl 4-bromo-2,5-bis(trifluoromethyl)benzoate 95%
Methyl 4-bromo-2,5-bis(trifluoromethyl)benzoate
Formel: C₁₀H₅BrF₆O₂
Molecular Weight: 351,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526891
CAS-Nummer: 1260793-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR