2-Chloro-5-(difluoromethoxy)pyrimidine 95%

Lieferant: Apollo Scientific
Achtung

PC56706-250MG PC56706-500MG
APOSPC56706-250MGEA 0 EUR
APOSPC56706-250MG APOSPC56706-500MG
2-Chloro-5-(difluoromethoxy)pyrimidine 95%
2-Chloro-5-(difluoromethoxy)pyrimidine
Formel: C₅H₃ClF₂N₂O
Molecular Weight: 180.54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415615
CAS-Nummer: 1192813-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR